Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP290 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536580
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536580 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536580 Supplier Invitrogen™ Supplier No. PA536580
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 290 kDa. Purity is >95% by SDS-PAGE.

This gene encodes a protein with 13 putative coiled-coil domains, a region with homology to SMC chromosome segregation ATPases, six KID motifs, three tropomyosin homology domains and an ATP/GTP binding site motif A. The protein is localized to the centrosome and cilia and has sites for N-glycosylation, tyrosine sulfation, phosphorylation, N-myristoylation, and amidation. Mutations in this gene have been associated with Joubert syndrome and nephronophthisis and the presence of antibodies against this protein is associated with several forms of cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP290
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Cep290
Gene Accession No. O15078, Q6A078
Gene Alias 3H11Ag; b2b1454Clo; b2b1752Clo; Bardet-Biedl syndrome 14 protein; Bardet-Biedl syndrome 14 protein homolog; BBS14; BC004690; Cancer/testis antigen 87; centrosomal protein 290; centrosomal protein 290kDa; centrosomal protein of 290 kDa; Cep290; CT87; CTCL tumor antigen se2-2; JBTS5; Kiaa0373; LCA10; Meckel syndrome, type 4; MKS4; monoclonal antibody 3H11 antigen; nephrocystin-6; nephrocytsin-6; Nphp6; POC3; POC3 centriolar protein homolog; prostate cancer antigen T21; rd16; RGD1311640; SLSN6; tumor antigen se2-2
Gene Symbols Cep290
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 781-830 of human CEP290.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216274, 314787, 80184
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.