Learn More
CEP290 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | CEP290 |
---|---|
Concentration | 0.51 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3874
![]() ![]() |
Proteintech Group Inc
224901AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Two isoforms were produced by alternative splicing with predicted MW of 290 and 180 kDa according to UniProt. 22490-1-AP can recognize both.
This gene encodes a protein with 13 putative coiled-coil domains, a region with homology to SMC chromosome segregation ATPases, six KID motifs, three tropomyosin homology domains and an ATP/GTP binding site motif A. The protein is localized to the centrosome and cilia and has sites for N-glycosylation, tyrosine sulfation, phosphorylation, N-myristoylation, and amidation. Mutations in this gene have been associated with Joubert syndrome and nephronophthisis and the presence of antibodies against this protein is associated with several forms of cancer.Specifications
CEP290 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O15078, Q6A078 | |
216274, 314787, 80184 | |
CEP290 Fusion Protein Ag17457 | |
Primary | |
-20°C | |
CEP290 |
0.51 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
3H11Ag, BBS14, Cancer/testis antigen 87, centrosomal protein 290kDa, Centrosomal protein of 290 kDa, CEP290, CT87, JBTS5, JBTS6, KIAA0373, LCA10, MKS4, Nephrocystin 6, rd16, SLSN6, Tumor antigen se2 2 | |
Cep290 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.