Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEP295 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558458
Description
Immunogen sequence: NTSSEQTGSS SFIPQLVQLS FTSLASAESG TILEPLFTES ESKIFSSHLQ IPQLQDRLLR ISQLIQPQQD NLKALQEQLA TQREAIILAR QEAREELL Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 67%.

Specifications
CEP295 | |
Polyclonal | |
Unconjugated | |
CEP295 | |
centriolar protein CEP295; centrosomal protein 295; centrosomal protein 295kDa; centrosomal protein of 295 kDa; CEP295; KIAA1731 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
85459 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9C0D2 | |
CEP295 | |
Recombinant protein corresponding to Human CEP295. Recombinant protein control fragment (Product #RP-102838). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction