Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP350 Antibody

Rabbit Polyclonal Antibody has been used in 6 publications
Supplier: Novus Biologicals NB10059811
Description
CEP350 Polyclonal specifically detects CEP350 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockdown Validated.Specifications
CEP350 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
CAP350GM133, centrosomal protein 350kDa, centrosome-associated protein 350, Centrosome-associated protein of 350 kDa, Cep350, FLJ38282, FLJ44058, KIAA0480centrosome associated protein 350 | |
Rabbit | |
351 kDa | |
100 μL | |
Primary | |
Human, Canine | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 2-5 ug/mg lysate | |
Q5VT06 | |
CEP350 | |
The immunogen recognized by this antibody maps to a region between residue 3066 and 3116 of human centrosome-associated protein 350 using the numbering given in entry NP_055625.3 (GeneID 9857). | |
Affinity Purified | |
RUO | |
9857 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction