Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEP350 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555826
Description
Immunogen sequence: ESEAKKLAGA SINYGSAWNT EYDVQQAPQE DGPWTKAVTP PVKDDNEDVF SARIQKMLGS CVSHATFDDD LPGVGNLSEF KKLPEMIRPQ SAISSFRVRS PGPK Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 74%.
May be involved in transcriptional regulation.
Specifications
| CEP350 | |
| Polyclonal | |
| Unconjugated | |
| CEP350 | |
| 4933409L06Rik; 6430546F08Rik; Cap350; centrosomal protein 350; centrosomal protein 350kDa; centrosome-associated protein 350; Centrosome-associated protein of 350 kDa; CEP350; GM133; Kiaa0480 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9857 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5VT06 | |
| CEP350 | |
| Recombinant protein corresponding to Human CEP350. Recombinant protein control fragment (Product #RP-94226). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction