Learn More
CEP41 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301798A
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a centrosomal and microtubule-binding protein which is predicted to have two coiled-coil domains and a rhodanese domain. In human retinal pigment epithelial cells the protein localized to centrioles and cilia. Mutations in this gene have been associated with Joubert Syndrome 15; an autosomal recessive ciliopathy and neurological disorder. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Mar 2012].Specifications
| CEP41 | |
| Polyclonal | |
| Unconjugated | |
| CEP41 | |
| JBTS15, TSGA14, centrosomal protein 41 kDa, centrosomal protein of 41 kDa, testis specific protein A14, testis specific, 14, testis-specific gene A14 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 95681 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q9BYV8 | |
| CEP41 | |
| Between 150 and 200 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.