Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP44 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559608
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559608 Supplier Invitrogen™ Supplier No. PA559608
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASCSDMDLLN PHRKSEVERP ASIPLSSGYS TASSDSTPRA STVNYCGLNE ISEETTIQKM ERMKKMFEET AELLKCPNHY Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 64%.

CEP44 (Centrosomal Protein 44) is a Protein Coding gene. Diseases associated with CEP44 include Retinitis Pigmentosa 29. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP44
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEP44
Gene Accession No. Q9C0F1
Gene Alias 4933440G23Rik; centrosomal protein 44; centrosomal protein 44kDa; centrosomal protein of 44 kDa; Cep44; HBV PreS1-transactivated protein 3; KIAA1712; mKIAA1712; PS1TP3
Gene Symbols CEP44
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CEP44. Recombinant protein control fragment (Product #RP-98216).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80817
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.