Learn More
Invitrogen™ CEP44 Polyclonal Antibody

Description
Immunogen sequence: VKVPEIKAEQ QDVNVNPEIT ALQTMLAECQ ENLKKLTSIE KRLDCLEQKM KGKVMVDENT WTNLLSRVTL LETEMLLSK Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 73%.
Specifications
Specifications
| Antigen | CEP44 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CEP44 |
| Gene Accession No. | Q9C0F1 |
| Gene Alias | 4933440G23Rik; centrosomal protein 44; centrosomal protein 44kDa; centrosomal protein of 44 kDa; Cep44; HBV PreS1-transactivated protein 3; KIAA1712; mKIAA1712; PS1TP3 |
| Gene Symbols | CEP44 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.