Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP55 Antibody (OTI10C4), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271990F
Description
CEP55 Monoclonal specifically detects CEP55 in Human samples. It is validated for Immunohistochemistry.Specifications
CEP55 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
CEP55 | |
Recombinant protein fragment corresponding to amino acids 1-280 of human CEP55 (NP_060601) produced in E.coli. | |
0.1 mL | |
55165.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI10C4 | |
Immunohistochemistry | |
C10orf3, cancer/testis antigen 111, centrosomal protein 55kDa, centrosomal protein of 55 kDa, Cep55, chromosome 10 open reading frame 3, CT111, FLJ10540, Up-regulated in colon cancer 6, URCC6 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction