Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP57 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106638
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106638 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106638 Supplier Invitrogen™ Supplier No. PA5106638
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CEP57.

Translokin binds basic fibroblast growth factor (FGF2; MIM 134920) and mediates its nuclear translocation and mitogenic activity (Bossard et al., 2003 [PubMed 12717444]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP57
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CEP57
Gene Accession No. Q86XR8, Q8CEE0
Gene Alias 3110002L15Rik; 4921510P06Rik; 4931428M20Rik; AI467480; centrosomal protein 57; centrosomal protein 57kDa; centrosomal protein of 57 kDa; Cep57; FGF2-interacting protein; KIAA0092; mKIAA0092; MVA2; PIG8; proliferation-inducing protein 8; RGD1309884; testis-specific protein 57; Translokin; Tsp57
Gene Symbols CEP57
Host Species Rabbit
Immunogen A synthesized peptide derived from human CEP57(Accession Q86XR8), corresponding to amino acid residues R242-V292.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74360, 9702
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.