Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP57L1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen CEP57L1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP170497
SDP
View Documents
Novus Biologicals
NBP170497
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

CEP57L1 Polyclonal specifically detects CEP57L1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CEP57L1
Polyclonal
Rabbit
centrosomal protein 57kDa-like 1, centrosomal protein 57kDa-like protein 1
CEP57L1
IgG
54 kDa
Western Blot
Unconjugated
RUO
285753
Synthetic peptides corresponding to C6ORF182 The peptide sequence was selected from the middle region of C6ORF182. Peptide sequence DIECELECLLKKMEIKGEQISKLKKHQDSVCKLQQKVQNSKMSEASGIQQ.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.