Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP57L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598613
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598613 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598613 Supplier Invitrogen™ Supplier No. PA598613
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CEP57L1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

C6orf182 also named as Centrosomal protein of 57 kDa related protein is a 460 amino-acid protein, which belongs to translokin family. C6orf182 as centrosomal protein may be required for microtubule attachment to centrosomes and localizes in the cytoplasm. The function of C6orf182 need to be further studied.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP57L1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CEP57L1
Gene Accession No. Q8IYX8
Gene Alias 2410017P07Rik; 4930484D11Rik; AI607327; bA487F23.2; C6orf182; centrosomal protein 57 like 1; centrosomal protein 57kDa-like 1; centrosomal protein 57kDa-like protein 1; centrosomal protein 57-like 1; Centrosomal protein CEP57L1; centrosomal protein of 57 kDa-related protein; CEP57L1; Cep57R; Cep57-related protein; RGD1311358
Gene Symbols CEP57L1
Host Species Rabbit
Immunogen Recombinant Human Centrosomal protein CEP57L1 protein (1-167AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285753
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.