Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEP57L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109515
Description
Immunogen sequence: MDSELMHSIV GSYHKPPERV FVPSFTQNEP SQNCHPANLE VTSPKILHSP NSQALILALK TLQEKIHRLE LERTQAEDNL NILSREAAQY KKALENETNE RNLAHQELIK QKKDISIQLS SAQSRCTLLE KQLEYTKRMV LNVEREKNMI LEQQAQLQRE KEQDQMK.
C6orf182 also named as Centrosomal protein of 57 kDa related protein is a 460 amino-acid protein, which belongs to translokin family. C6orf182 as centrosomal protein may be required for microtubule attachment to centrosomes and localizes in the cytoplasm. The function of C6orf182 need to be further studied.
Specifications
CEP57L1 | |
Polyclonal | |
Unconjugated | |
CEP57L1 | |
2410017P07Rik; 4930484D11Rik; AI607327; bA487F23.2; C6orf182; centrosomal protein 57 like 1; centrosomal protein 57kDa-like 1; centrosomal protein 57kDa-like protein 1; centrosomal protein 57-like 1; Centrosomal protein CEP57L1; centrosomal protein of 57 kDa-related protein; CEP57L1; Cep57R; Cep57-related protein; RGD1311358 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
285753 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IYX8 | |
CEP57L1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-167 of human CEP57L1 (NP_776191.1). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction