Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEP57L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559153
Description
Immunogen sequence: SKKTKCIKRR PPWQICSKFG ALPFVAEKMR QHRDPHILQK PFNVTETRCL PKPSRTTSWC KAIPPDSEKS ISICDNLSEL LMAMQDELDQ MSMEHQE Highest antigen sequence identity to the following orthologs: Mouse - 44%, Rat - 43%.
C6orf182 also named as Centrosomal protein of 57 kDa related protein is a 460 amino-acid protein, which belongs to translokin family. C6orf182 as centrosomal protein may be required for microtubule attachment to centrosomes and localizes in the cytoplasm. The function of C6orf182 need to be further studied.
Specifications
CEP57L1 | |
Polyclonal | |
Unconjugated | |
CEP57L1 | |
2410017P07Rik; 4930484D11Rik; AI607327; bA487F23.2; C6orf182; centrosomal protein 57 like 1; centrosomal protein 57kDa-like 1; centrosomal protein 57kDa-like protein 1; centrosomal protein 57-like 1; Centrosomal protein CEP57L1; centrosomal protein of 57 kDa-related protein; CEP57L1; Cep57R; Cep57-related protein; RGD1311358 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
285753 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IYX8 | |
CEP57L1 | |
Recombinant protein corresponding to Human CEP57L1. Recombinant protein control fragment (Product #RP-95018). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction