Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP63 Antibody (OTI2H2), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272221C
Description
CEP63 Monoclonal specifically detects CEP63 in Human samples. It is validated for Western Blot.Specifications
CEP63 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CEP63 | |
Full length recombinant protein of human CEP63 (NP_001035859) produced in E.coli. | |
0.1 mL | |
80254.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI2H2 | |
Western Blot | |
centrosomal protein 63kDa, centrosomal protein of 63 kDa, centrosome protein CEP63, Cep63, FLJ13386, MGC78416 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction