Learn More
CEP63 Mouse anti-Human, Clone: OTI2F8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809290

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CEP63 is a protein with six coiled-coil domains. The protein is localized to the centrosome, a non-membraneous organelle that functions as the major microtubule-organizing center in animal cells.Specifications
CEP63 | |
Monoclonal | |
Unconjugated | |
CEP63 | |
SCKL6 | |
Mouse | |
Affinity Chromatography | |
RUO | |
80254 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2F8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96MT8 | |
CEP63 | |
Full length human recombit protein of human CEP63 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.