Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEP63 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103728
Description
Antibody detects endogenous levels of total CEP63.
CEP63 is a protein with six coiled-coil domains. The protein is localized to the centrosome, a non-membranous organelle that functions as the major microtubule-organizing center in animal cells.
Specifications
CEP63 | |
Polyclonal | |
Unconjugated | |
CEP63 | |
4921501M07; AL450317.13gm1; AW107703; CD20R; centrosomal protein 63; centrosomal protein 63kDa; centrosomal protein of 63 kDa; centrosome protein Cep63; CEP63; D9Mgc41; D9Mgc48e; ET2; LOW QUALITY PROTEIN: centrosomal protein of 63 kDa; SCKL6 | |
Rabbit | |
Affinity chromatography | |
RUO | |
28135, 80254 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q3UPP8, Q96MT8 | |
CEP63 | |
A synthesized peptide derived from human CEP63(Accession Q96MT8), corresponding to amino acid residues D539-I589. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction