Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP63 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109516
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109516 Supplier Invitrogen™ Supplier No. PA5109516
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEALLEGIQN RGHGGGFLTS CEAELQELMK QIDIMVAHKK SEWEGRTHAL ETCLKIREQE LKSLRSQLDV THKEVGMLHQ QVEEHEKIKQ EMTMEYKQEL KKLHEELCIL KRSYEKLQKK QMREFRGNTK NHREDRSEIE RLTAKIEEFR QKSLDWEKQR LIYQQQVSSL EAQRKALAEQ SEIIQAQLVN RKQKLESVEL SSQSEIQHLS SKLERANDTI CANELEIERL TMRVNDLVGT SMTVLQEQQQ KEEKLRESEK LLEALQEEKR ELKAALQSQE NLIHEARIQK EKLQEKVKAT.

CEP63 is a protein with six coiled-coil domains. The protein is localized to the centrosome, a non-membranous organelle that functions as the major microtubule-organizing center in animal cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP63
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CEP63
Gene Accession No. Q3UPP8, Q4KLY0, Q96MT8
Gene Alias 4921501M07; AL450317.13gm1; AW107703; CD20R; centrosomal protein 63; centrosomal protein 63kDa; centrosomal protein of 63 kDa; centrosome protein Cep63; CEP63; D9Mgc41; D9Mgc48e; ET2; LOW QUALITY PROTEIN: centrosomal protein of 63 kDa; SCKL6
Gene Symbols CEP63
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human CEP63 (NP_001035859.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28135, 300963, 80254
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.