Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP68 Antibody (OTI5B4), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271728UV
Description
CEP68 Monoclonal specifically detects CEP68 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CEP68 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CEP68 | |
Full length recombinant protein of human CEP68 (NP_055962) produced in HEK293T cells. | |
0.1 mL | |
23177.0 | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI5B4 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
centrosomal protein 68kDa, centrosomal protein of 68 kDa, Cep68, FLJ36750, KIAA0582FLJ25920 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction