Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP68 Monoclonal Antibody (OTI5B4)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526303

Catalog No. PIMA526303


Only null left
Add to Cart

Description

Description

CEP68 Monoclonal Antibody for Western Blot, ICC/IF

Centrosomal protein of 68 kDa is a protein that in humans is encoded by the CEP68 gene. It is mapped to chromosome 2. CEP68 is required for centrosome cohesion. It decorates fibres emanating from the proximal ends of centrioles. CEP68 and rootletin depend both on each other for centriole association, and both also require CEP250 for their function.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CEP68
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q76N32
CEP68
Full length human recombinant protein of CEP68 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Western Blot, Immunocytochemistry
OTI5B4
Unconjugated
CEP68
6030463E10Rik; AI481761; BC027174; centrosomal protein 68; centrosomal protein 68kDa; centrosomal protein of 68 kDa; Cep68; Kiaa0582; RGD1309101
Mouse
Affinity chromatography
RUO
23177
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.