Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP68 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559227
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559227 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559227 Supplier Invitrogen™ Supplier No. PA559227
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSPQLRTRDR GWPSPRPERE KRTSQSARRP TCTESRWKSE EEVESDDEYL ALPARLTQVS SLVSYLGSIS TLVTLPT Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 62%.

Centrosomal protein of 68 kDa is a protein that in humans is encoded by the CEP68 gene. It is mapped to chromosome 2. CEP68 is required for centrosome cohesion. It decorates fibres emanating from the proximal ends of centrioles. CEP68 and rootletin depend both on each other for centriole association, and both also require CEP250 for their function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP68
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEP68
Gene Accession No. Q76N32
Gene Alias 6030463E10Rik; AI481761; BC027174; centrosomal protein 68; centrosomal protein 68kDa; centrosomal protein of 68 kDa; Cep68; Kiaa0582; RGD1309101
Gene Symbols CEP68
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CEP68. Recombinant protein control fragment (Product #RP-98199).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23177
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.