Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP70 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983154
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3154 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3154 Supplier Bioss Supplier No. BS13860R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CEP70 plays a role in the organization of both preexisting and nascent microtubules in interphase cells. During mitosis, it is required for the organization and orientation of the mitotic spindle.

Specifications

Antigen CEP70
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CEP70
Gene Accession No. Q8NHQ1
Gene Alias 6720484E09Rik; BITE; C030018L16Rik; centrosomal protein 70; centrosomal protein 70kDa; Centrosomal protein of 70 kDa; Cep70; p10-binding protein; testicular tissue protein Li 36
Gene Symbols CEP70
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CEP70.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80321
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.