Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP72 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB246598 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB246598 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB246598 Supplier Novus Biologicals Supplier No. NB10060662AF700
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CEP72 Polyclonal antibody specifically detects CEP72 in Human samples. It is validated for Western Blot

Specifications

Antigen CEP72
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias centrosomal protein 72kDa, centrosomal protein of 72 kDa, Cep72, KIAA1519FLJ10565, MGC5307
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 250 and 300 of centrosomal protein 72kDa using the numbering given in entry NP_060610.2 (GeneID 55722).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55722
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.