Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP72 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060662R
Description
CEP72 Polyclonal antibody specifically detects CEP72 in Human samples. It is validated for Western BlotSpecifications
CEP72 | |
Polyclonal | |
Western Blot | |
centrosomal protein 72kDa, centrosomal protein of 72 kDa, Cep72, KIAA1519FLJ10565, MGC5307 | |
The immunogen recognized by this antibody maps to a region between residue 250 and 300 of centrosomal protein 72kDa using the numbering given in entry NP_060610.2 (GeneID 55722). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55722 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction