Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP72 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060661JF646
Description
CEP72 Polyclonal antibody specifically detects CEP72 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)Specifications
CEP72 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
centrosomal protein 72kDa, centrosomal protein of 72 kDa, Cep72, KIAA1519FLJ10565, MGC5307 | |
The immunogen recognized by this antibody maps to a region between residue 150 and 200 of centrosomal protein 72kDa using the numbering given in entry NP_060610.2 (GeneID 55722). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55722 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction