Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEP72 Monoclonal Antibody (OTI4D2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527404
Description
CEP72 Monoclonal Antibody for Western Blot, IHC (P)
Involved in the recruitment of key centrosomal proteins to the centrosome. Provides centrosomal microtubule-nucleation activity on the gamma-tubulin ring complexes (gamma-TuRCs) and has critical roles in forming a focused bipolar spindle, which is needed for proper tension generation between sister chromatids. Required for localization of KIZ, AKAP9 and gamma-tubulin ring complexes (gamma-TuRCs).
Specifications
CEP72 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9P209 | |
CEP72 | |
Full length human recombinant protein of CEP72 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4D2 | |
Unconjugated | |
CEP72 | |
2610029E11Rik; 4933440J22Rik; centrosomal protein 72; centrosomal protein 72 kDa; centrosomal protein 72kDa; centrosomal protein of 72 kDa; Cep72; KIAA1519 | |
Mouse | |
Affinity chromatography | |
RUO | |
55722 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction