Learn More
CEP72 Mouse anti-Human, Clone: OTI1A5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811115

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Involved in the recruitment of key centrosomal proteins to the centrosome. Provides centrosomal microtubule-nucleation activity on the gamma-tubulin ring complexes (gamma-TuRCs) and has critical roles in forming a focused bipolar spindle, which is needed for proper tension generation between sister chromatids. Required for localization of KIZ, AKAP9 and gamma-tubulin ring complexes (gamma-TuRCs).Specifications
CEP72 | |
Monoclonal | |
Unconjugated | |
CEP72 | |
centrosomal protein 72 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55722 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1A5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9P209 | |
CEP72 | |
Full length human recombit protein of human CEP72 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.