Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP72 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563537
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563537 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563537 Supplier Invitrogen™ Supplier No. PA563537
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YQCGDSGKQG RETRRSSCRG CCLEKMPWSQ LCGELPPLYG AEPEASRAPR PHTYFTPHPD SMDTEDSASS QKLDLSGEMV PGPLPAPGKC RKRRMP Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 48%.

Involved in the recruitment of key centrosomal proteins to the centrosome. Provides centrosomal microtubule-nucleation activity on the gamma-tubulin ring complexes (gamma-TuRCs) and has critical roles in forming a focused bipolar spindle, which is needed for proper tension generation between sister chromatids. Required for localization of KIZ, AKAP9 and gamma-tubulin ring complexes (gamma-TuRCs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP72
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEP72
Gene Accession No. Q9P209
Gene Alias 2610029E11Rik; 4933440J22Rik; centrosomal protein 72; centrosomal protein 72 kDa; centrosomal protein 72kDa; centrosomal protein of 72 kDa; Cep72; KIAA1519
Gene Symbols CEP72
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CEP72. Recombinant protein control fragment (Product #RP-96867).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55722
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.