Learn More
CEP72 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301298AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Involved in the recruitment of key centrosomal proteins to the centrosome. Provides centrosomal microtubule-nucleation activity on the gamma-tubulin ring complexes (gamma-TuRCs) and has critical roles in forming a focused bipolar spindle, which is needed for proper tension generation between sister chromatids. Required for localization of KIZ, AKAP9 and gamma-tubulin ring complexes (gamma-TuRCs).Specifications
| CEP72 | |
| Polyclonal | |
| Unconjugated | |
| CEP72 | |
| Centrosomal protein of 72 kDa, Cep72, CEP72, KIAA1519 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55722 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q9P209 | |
| CEP72 | |
| Between 250 and 300 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.