Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP76 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128749AF532
Description
CEP76 Polyclonal antibody specifically detects CEP76 in Human samples. It is validated for Western BlotSpecifications
CEP76 | |
Polyclonal | |
Western Blot | |
C18orf9, centrosomal protein 76kDa, centrosomal protein of 76 kDa, Cep76, chromosome 18 open reading frame 9, FLJ12542, HsT1705 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 30 of human centrosomal protein 76kDa using the numbering given in entry NP_079175.2 (GeneID 79959). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
79959 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction