Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP76 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558772
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558772 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558772 Supplier Invitrogen™ Supplier No. PA558772
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTNPDEPPVA EQPKPLYPYR TIGCVFNHQM FLGNCQPSDA VETCVFDLND ESKWKPMSEE AIKSVCAPGA TTSLPPFPPL CASTIDASVT SNEIEMQLRL LVSEHRKDLG LTTVWED Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

This gene encodes a centrosomal protein which regulates centriole amplification by limiting centriole duplication to once per cell cycle. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Dec 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP76
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEP76
Gene Accession No. Q8TAP6
Gene Alias 6230425F05Rik; 9630013E05; C18orf9; centrosomal protein 76; centrosomal protein 76kDa; centrosomal protein of 76 kDa; Cep76; CSG11; HsT1705; RGD1305183
Gene Symbols CEP76
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CEP76. Recombinant protein control fragment (Product #RP-98210).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79959
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.