Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP76 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113290
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113290 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113290 Supplier Invitrogen™ Supplier No. PA5113290
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CEP76 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a centrosomal protein which regulates centriole amplification by limiting centriole duplication to once per cell cycle. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Dec 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP76
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CEP76
Gene Accession No. Q8TAP6
Gene Alias 6230425F05Rik; 9630013E05; C18orf9; centrosomal protein 76; centrosomal protein 76kDa; centrosomal protein of 76 kDa; Cep76; CSG11; HsT1705; RGD1305183
Gene Symbols CEP76
Host Species Rabbit
Immunogen Recombinant Human Centrosomal protein of 76 kDa protein (241-550AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79959
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.