Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP76 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101511
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101511 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101511 Supplier Invitrogen™ Supplier No. PA5101511
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CEP76.

This gene encodes a centrosomal protein which regulates centriole amplification by limiting centriole duplication to once per cell cycle. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Dec 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP76
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CEP76
Gene Accession No. Q0VEJ0, Q8TAP6
Gene Alias 6230425F05Rik; 9630013E05; C18orf9; centrosomal protein 76; centrosomal protein 76kDa; centrosomal protein of 76 kDa; Cep76; CSG11; HsT1705; RGD1305183
Gene Symbols CEP76
Host Species Rabbit
Immunogen A synthesized peptide derived from human CEP76(Accession Q8TAP6), corresponding to amino acid residues E251-H301.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225659, 291540, 79959
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.