Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP83 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121204
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121204 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121204 Supplier Invitrogen™ Supplier No. PA5121204
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HT-29, Mouse lung, Mouse spleen, Mouse thymus. The target is usually found in the following locations: Cytoplasm, centriole, centrosome, cytoskeleton, microtubule organizing center. Immunogen sequence: MLIDERLRCE HHKANYQTLK AEHTRLQNEH VKLQNELKHL FNEKQTQQEK LQLLLEELRG ELVEKTKDLE EMKLQILTPQ KLELLRAQIQ QELETPMRER FRNLDEEVEK YRAVYNKLRY EHTFLKSEFE HQKEEYARIL DEGKIKYESE IARLEEDKEE LRNQLLNVDL TKDSKRVEQL AREKVYLCQK LKGLEAEVAE LKAEKENSEA QVENAQRIQV RQLAEMQ.

Component of the distal appendage region of the centriole involved in the initiation of primary cilium assembly. May collaborate with IFT20 in the trafficking of ciliary membrane proteins from the Golgi complex to the cilium during the initiation of primary cilium assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP83
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CEP83
Gene Accession No. Q9D5R3, Q9Y592
Gene Alias 2600001G24Rik; 4921537D05Rik; 5730513H21Rik; CCDC41; centrosomal protein 83; centrosomal protein 83kDa; centrosomal protein of 83 kDa; Cep83; coiled-coil domain containing 41; coiled-coil domain-containing protein 41; NPHP18; NY-REN-58; NY-REN-58 antigen; renal carcinoma antigen NY-REN-58
Gene Symbols CEP83
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 34-260 of human CEP83 (NP_057206.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51134, 77048
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.