Learn More
CEP85 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303326AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that belongs to the centrosome-associated family of proteins. The centrosome is a subcellular organelle in the animal cell that functions as a microtubule organizing center and is involved in cell-cycle progression. Alternate splicing results in multiple transcript variants.Specifications
CEP85 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q6P2H3 | |
CEP85 | |
Between 675 and 725 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CEP85 | |
CCDC21, centrosomal protein of 85 kDa, coiled-coil domain containing 21, coiled-coil domain-containing protein 21 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64793 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.