Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEP85L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556070
Description
Immunogen sequence: SKLRESLTPD GSKWSTSLMQ TLGNHSRGEQ DSSLDMKDFR PLRKWSSLSK LTAPDNCGQG GTVCREESRN GLEK Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 68%.
The protein encoded by this gene was identified as a breast cancer antigen. Nothing more is known of its function at this time. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
CEP85L | |
Polyclonal | |
Unconjugated | |
CEP85L | |
bA57K17.2; C1H6orf204; C6orf204; centrosomal protein 85 like; centrosomal protein 85kDa-like; centrosomal protein 85-like; centrosomal protein of 85 kDa-like; CEP85L; coiled-coil domain-containing c6orf204-like protein; ENSMUSG00000038594; Gm9766; LOW QUALITY PROTEIN: centrosomal protein of 85 kDa-like; NY-BR-15; rCG22129-like; serologically defined breast cancer antigen NY-BR-15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
387119 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5SZL2 | |
CEP85L | |
Recombinant protein corresponding to Human CEP85L. Recombinant protein control fragment (Product #RP-94648). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction