Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP85L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556071
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556071 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556071 Supplier Invitrogen™ Supplier No. PA556071
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDCKYKFESC SKEDFRASSS TLRRQPVDMT YSALPESKPI MTSSEAFEPP KYLMLGQQAV GGVPIQPSVR TQMW Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 82%.

The protein encoded by this gene was identified as a breast cancer antigen. Nothing more is known of its function at this time. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP85L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEP85L
Gene Accession No. Q5SZL2
Gene Alias bA57K17.2; C1H6orf204; C6orf204; centrosomal protein 85 like; centrosomal protein 85kDa-like; centrosomal protein 85-like; centrosomal protein of 85 kDa-like; CEP85L; coiled-coil domain-containing c6orf204-like protein; ENSMUSG00000038594; Gm9766; LOW QUALITY PROTEIN: centrosomal protein of 85 kDa-like; NY-BR-15; rCG22129-like; serologically defined breast cancer antigen NY-BR-15
Gene Symbols CEP85L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CEP85L. Recombinant protein control fragment (Product #RP-94649).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387119
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.