Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP95 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562608
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562608 Supplier Invitrogen™ Supplier No. PA562608
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTIANNLLFK CHIHLRIHEL QDCDANVFIA LYQSILGEKV PDLIVIPRSQ EDDAHNVQAV IDSLALDYLQ VSLSHITGEN IVKGDKESIK NLLEIFDGLL EYLT Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 97%.

CEP95 gene ontology (GO) annotation includes protein binding and cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP95
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEP95
Gene Accession No. Q96GE4
Gene Alias 4732496G21Rik; AI118346; AI448335; Ccdc45; centrosomal protein 95; centrosomal protein 95kDa; centrosomal protein of 95 kDa; CEP45; CEP95; coiled-coil domain containing 45; coiled-coil domain-containing protein 45; D330027A14Rik; F630025I20Rik; RGD1309655
Gene Symbols CEP95
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CEP95. Recombinant protein control fragment (Product #RP-104247).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90799
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.