Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP95 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101534
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101534 Supplier Invitrogen™ Supplier No. PA5101534
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CCDC45.

CEP95 gene ontology (GO) annotation includes protein binding and cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP95
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CEP95
Gene Accession No. Q5XI03, Q8BVV7, Q96GE4
Gene Alias 4732496G21Rik; AI118346; AI448335; Ccdc45; centrosomal protein 95; centrosomal protein 95kDa; centrosomal protein of 95 kDa; CEP45; CEP95; coiled-coil domain containing 45; coiled-coil domain-containing protein 45; D330027A14Rik; F630025I20Rik; RGD1309655
Gene Symbols CEP95
Host Species Rabbit
Immunogen A synthesized peptide derived from human CEP95(Accession Q96GE4), corresponding to amino acid residues Q590-Q640.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287766, 320162, 90799
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.