Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP97 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP106508G
Description
CEP97 Polyclonal antibody specifically detects CEP97 in Human samples. It is validated for Western BlotSpecifications
CEP97 | |
Polyclonal | |
Western Blot | |
2810403B08Rik, centrosomal protein 97kDa, centrosomal protein of 97 kDa, Cep97, FLJ23047, FLJ26462, Leucine-rich repeat and IQ domain-containing protein 2, leucine-rich repeats and IQ motif containing 2, LRRIQ2 | |
The immunogen recognized by this antibody maps to a region between residue 625 and 675 of human centrosomal protein 97kDa using the numbering given in entry NP_078824.2 (GeneID 79598). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
79598 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction