Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP97 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551790
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551790 Supplier Invitrogen™ Supplier No. PA551790
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QEEAFRFLWN QVRSLQVWQQ TVDQRLSSWH TDVPPISSTL VPSKHPLFTQ SQESSCDQNA DWFIASDVAP QEKSLPEFPD SGFHSSLTEQ VHSLQHSLDF EKSSTEGSES SIMGNSIDTV RYGKESDLGD VSEEHGEWN Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 68%.

CEP97 (centrosomal protein of 97 kDa) has been identified as part of a complex with CP110. Together, CP110 and CEP97 have been shown to play a role in the assembly of cilia. Loss of CEP97 or CEP110 promotes cilia formation while forced expression inhibits ciliogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP97
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEP97
Gene Accession No. Q8IW35
Gene Alias 2810403B08Rik; 4932439K18Rik; AA437768; centrosomal protein 97; centrosomal protein 97kDa; centrosomal protein of 97 kDa; Cep97; E130116N02Rik; Leucine-rich repeat and IQ domain-containing protein 2; leucine-rich repeats and IQ motif containing 2; LRRIQ2
Gene Symbols CEP97
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CEP97. Recombinant protein control fragment (Product #RP-94776).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79598
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.