Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Cepheid Xpert™ GBS LB Control Panel
SDP

Catalog No. 23017298
Click to view available options
Quantity:
12 Swabs
This item is not returnable. View return policy
This item is not returnable. View return policy

Microbiologics offers a wide range of bundled QC Sets and Panels for the most commonly used technologies and standards. Each set or panel contains the manufacturer-recommended quality control strains for a particular instrument or test method in a simple, ready-to-use format.

  • Cepheid Xpert™ GBS LB Control Panel features the manufacturer recommended QC strains in an easy-to-use, inactivated Helix Elite format.
  • Each kit includes 6 positive control swabs, 6 negative control swabs, and a Certificate of Analysis
  • Positive control includes Streptococcus agalactiae
  • Negative control includes Lactobacillus acidophilus.

Specifications

Biosafety Level 1
Content And Storage 2°C to 8°C
Format QC Sets and Panels: Helix Elite™
Manufacturing Origin United States
Product Type Microbiology Media
Packaging Type Plastic Box
Quantity 12 Swabs
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.