Learn More
Invitrogen™ CER1 Polyclonal Antibody

Description
Cerberus 1 Polyclonal Antibody detects endogenous levels of Cerberus 1 protein.
Specifications
Specifications
| Antigen | CER1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | CER1 |
| Gene Accession No. | O95813 |
| Gene Alias | Cer1; cer-1; Cerberus; cerberus 1 homolog (Xenopus laevis); cerberus 1, cysteine knot superfamily, homolog; cerberus 1, DAN family BMP antagonist; cerberus homolog; cerberus-like protein; cerberus-related 1; cerberus-related protein; Cerl; Cer-l; Cerl1; Cerr1; DAN domain family member 4; DAND4; MGC119894; MGC119895; MGC96951; RGD1563046 |
| Gene Symbols | CER1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.