Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CER1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578286
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578286 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578286 Supplier Invitrogen™ Supplier No. PA578286
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted Reactivity: Rhesus Monkey (89%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a cytokine member of the cysteine knot superfamily, characterized by nine conserved cysteines and a cysteine knot region. The cerberus-related cytokines, together with Dan and DRM/Gremlin, represent a group of bone morphogenetic protein antagonists that can bind directly to BMPs and inhibit their activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CER1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene CER1
Gene Accession No. O95813
Gene Alias Cer1; cer-1; Cerberus; cerberus 1 homolog (Xenopus laevis); cerberus 1, cysteine knot superfamily, homolog; cerberus 1, DAN family BMP antagonist; cerberus homolog; cerberus-like protein; cerberus-related 1; cerberus-related protein; Cerl; Cer-l; Cerl1; Cerr1; DAN domain family member 4; DAND4; MGC119894; MGC119895; MGC96951; RGD1563046
Gene Symbols CER1
Host Species Rabbit
Immunogen Full length human CER1 Recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9350
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.