Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceramide glucosyltransferase Polyclonal Antibody, Cyanine7, Bioss
SDP

Catalog No. 502280892
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-0892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-0892 Supplier Bioss Supplier No. BS21562RCY7
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Ceramide glucosyltransferase Polyclonal antibody specifically detects Ceramide glucosyltransferase in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)

Specifications

Antigen Ceramide glucosyltransferase
Applications Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Cyanine7
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene UGCG
Gene Accession No. O88693, Q16739
Gene Alias AU043821; C80537; cb539; Ceramide glucosyltransferase; ectoplacental cone, invasive trophoblast giant cells, extraembryonic ectoderm and chorion sequence 21; Epcs21; GCS; GlcT; GLCT1; GLCT-1; glucosylceramide synthase; hypothetical protein LOC553944; sb:cb539; UDP-glucose ceramide glucosyltransferase; UDP-glucose:ceramide glycosyltransferase; UDP-glucose:N-acylsphingosine D-glucosyltransferase; Ugcg; Ugcgl; zgc:112506
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Ceramide glucosyltransferase
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22234, 7357
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.