Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ceramide Kinase Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002911H
Description
Ceramide Kinase Polyclonal specifically detects Ceramide Kinase in Human, Mouse samples. It is validated for Western Blot.Specifications
Ceramide Kinase | |
Polyclonal | |
Western Blot | |
Q8TCT0 | |
CERK | |
A synthetic peptide made to an internal portion of human Ceramide Kinase (between residues 50-150). [Swiss-Prot# Q8TCT0] | |
Affinity Purified | |
RUO | |
Primary | |
Human. Immunogen has 86% homology to mouse. | |
Store at 4C in the dark. |
Western Blot | |
HRP | |
PBS with No Preservative | |
ceramide kinase, dA59H18.2, DKFZp434E0211, EC 2.7.1.138, FLJ21430, FLJ23239, hCERKLipid kinase 4, KIAA1646dA59H18.3, lipid kinase LK4, LK4Acylsphingosine kinase, MGC131878 | |
Rabbit | |
65 kDa | |
0.1 mL | |
Apoptosis, Cell Biology, Lipid and Metabolism | |
64781 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction