Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cerberus 1 Antibody (034), Alexa Fluor™ 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289662AF405
Description
Cerberus 1 Monoclonal antibody specifically detects Cerberus 1 in Human samples. It is validated for ELISASpecifications
Cerberus 1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
034 | |
ELISA | |
cerberus, cerberus 1 (Xenopus laevis) homolog (cysteine knot superfamily), cerberus 1, cysteine knot superfamily, homolog (Xenopus laevis), cerberus-related 1, Cerberus-related protein, DAN domain family member 4, DAND4, MGC119894, MGC119895, MGC96951 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cerberus 1 (AAH69371.1; Met1-Ala267). | |
0.1 mL | |
Stem Cell Markers, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
9350 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction