Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cerberus 1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299621MFV450
Description
Cerberus 1 Polyclonal antibody specifically detects Cerberus 1 in Human samples. It is validated for ELISASpecifications
Cerberus 1 | |
Polyclonal | |
ELISA | |
cerberus, cerberus 1 (Xenopus laevis) homolog (cysteine knot superfamily), cerberus 1, cysteine knot superfamily, homolog (Xenopus laevis), cerberus-related 1, Cerberus-related protein, DAN domain family member 4, DAND4, MGC119894, MGC119895, MGC96951 | |
Produced in rabbits immunized with purified, recombinant Human Cerberus 1 (Accession#: AAH69371.1; Met1-Ala267) | |
0.1 mL | |
Stem Cell Markers, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
9350 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction