Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CERCAM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532118
Description
Recommended positive controls: GL261, C8D30, NIH-3T3, mouse brain. Predicted reactivity: Mouse (83%), Rat (86%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Probable cell adhesion protein involved in leukocyte transmigration across the blood-brain barrier. Does not express any beta-galactosyltransferase activity in vitro. [UniProt].
Specifications
CERCAM | |
Polyclonal | |
Unconjugated | |
CERCAM | |
2600006K01Rik; AL024097; CEECAM1; CERCAM; cerebral cell adhesion molecule; cerebral endothelial cell adhesion molecule; cerebral endothelial cell adhesion molecule 1; GLT25D3; glycosyltransferase 25 domain containing 3; glycosyltransferase 25 family member 3; Inactive glycosyltransferase 25 family member 3; KIAA1502; Probable inactive glycosyltransferase 25 family member 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
296616, 51148, 99151 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
A3KGW5, Q5T4B2, Q5U309 | |
CERCAM | |
Recombinant protein fragment corresponding to a region within amino acids 14 and 234 of Human CERCAM. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction