Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CERCAM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554487
Description
Immunogen sequence: TFLASLRAEG ADQLAFYPPH PNYTWPFDDI IVFAYACQAA GVSVHVCNEH RYGYMNVPVK SHQGLEDERV NFIHLILEAL VDG Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 84%.
Probable cell adhesion protein involved in leukocyte transmigration across the blood-brain barrier. Does not express any beta-galactosyltransferase activity in vitro. [UniProt].
Specifications
CERCAM | |
Polyclonal | |
Unconjugated | |
CERCAM | |
2600006K01Rik; AL024097; CEECAM1; CERCAM; cerebral cell adhesion molecule; cerebral endothelial cell adhesion molecule; cerebral endothelial cell adhesion molecule 1; GLT25D3; glycosyltransferase 25 domain containing 3; glycosyltransferase 25 family member 3; Inactive glycosyltransferase 25 family member 3; KIAA1502; Probable inactive glycosyltransferase 25 family member 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51148 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T4B2 | |
CERCAM | |
Recombinant protein corresponding to Human CERCAM. Recombinant protein control fragment (Product #RP-93040). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction